Learn More
Medchemexpress LLC Tetrahydro-4H-pyran-4-one | 29943-42-8 | 99.9% | C5H8O2 | 50 G
SDP
Supplier:  Medchemexpress LLC HY7648750G
Encompass_Preferred
Tetrahydro-4H-pyran-4-one is a biochemical reagent suitable for life science research as a biological material or organic compound. It is used in the preparation of 4-methoxytetrahydropyran-4-yl protecting groups and the synthesis of symmetric tetra substituted methanes.
  • Methyl enol ether form serves as a protecting agent for alcohols in nucleotide synthesis.
  • Applied in studies involving enantioselective alpha-aminoxylation of ketones.
  • Participates in condensation reactions for dipeptide and spiroimidazolone preparation.
  • Used in Wittig reactions for penicillin synthesis.
  • Utilized in a ring of vitamin D3.
Catalog No. 50-004-34520