Learn More
Medchemexpress LLC Tetrahydro-4H-pyran-4-one | 29943-42-8 | MFCD00006581 | 100.0% | C5H8O2 | 500 G
SDP
Catalog No. 5000433856
Encompass_Preferred
Tetrahydro-4H-pyran-4-one is a biochemical reagent suitable for life science-related research as a biological material or organic compound.
  • Used in the preparation of 4-methoxytetrahydropyran-4-yl protecting group.
  • Employed in the synthesis of symmetric tetra-substituted methanes.
  • Functions as a protecting agent for alcohols, particularly in nucleotide synthesis.
  • Utilized in a study focusing on the enantioselective alpha-aminoxylation of ketones.
  • Undergoes condensation reactions for the preparation of dipeptides and spiroimidazolones.
  • Employed in Wittig reactions for the synthesis of penicillins and in a ring of vitamin D3.
Catalog No. 50-004-33856 Supplier Medchemexpress LLC Supplier No. HY76487500G
Tetrahydro-4H-pyran-4-one is a biochemical reagent suitable for life science-related research as a biological material or organic compound.
  • Used in the preparation of 4-methoxytetrahydropyran-4-yl protecting group.
  • Employed in the synthesis of symmetric tetra-substituted methanes.
  • Functions as a protecting agent for alcohols, particularly in nucleotide synthesis.
  • Utilized in a study focusing on the enantioselective alpha-aminoxylation of ketones.
  • Undergoes condensation reactions for the preparation of dipeptides and spiroimidazolones.
  • Employed in Wittig reactions for the synthesis of penicillins and in a ring of vitamin D3.