Learn More
Apexbio Technology LLC Tetrahydro-β-carboline 16502-01-5 5g
SDP
Catalog No. 5000097168
Encompass_Preferred
Tetrahydro- -carboline (CAS 16502-01-5) is a -carboline alkaloid derivative characterized by a tetrahydroindole fused to a pyridine ring This compound modulates neurotransmitter dynamics in the central nervous system by interacting with specific receptors and enzymes thereby influencing neurotransmitter release and metabolism Tetrahydro- -carboline has been utilized in studies on drug dependence neuroprotection and cognitive function within neuropharmacology Its molecular framework supports applications in the development of lead compounds and the investigation of neurobiological pathways involved in central nervous system disorders
Catalog No. 50-000-97168 Supplier Apexbio Technology LLC Supplier No. A47115000
Tetrahydro- -carboline (CAS 16502-01-5) is a -carboline alkaloid derivative characterized by a tetrahydroindole fused to a pyridine ring This compound modulates neurotransmitter dynamics in the central nervous system by interacting with specific receptors and enzymes thereby influencing neurotransmitter release and metabolism Tetrahydro- -carboline has been utilized in studies on drug dependence neuroprotection and cognitive function within neuropharmacology Its molecular framework supports applications in the development of lead compounds and the investigation of neurobiological pathways involved in central nervous system disorders