Learn More
Medchemexpress LLC Tetrahydro-β-carboline | 16502-01-5 | 99.4% | 172.23 | 10 MM * 1 ML
SDP
Catalog No. 5000401336
Encompass_Preferred
Tetrahydro-β-carboline (Tryptoline) is a metabolite of tryptamine and a competitive serotonin reuptake inhibitor. It is intended for research use only and not for patient use.
  • Metabolite of tryptamine
  • Competitive serotonin reuptake inhibitor with a Ki value of 6.1 μM
  • Increases serotonin levels in the brain of rats in vivo studies
Catalog No. 50-004-01336 Supplier Medchemexpress LLC Supplier No. HY2069610MM/1ML
Tetrahydro-β-carboline (Tryptoline) is a metabolite of tryptamine and a competitive serotonin reuptake inhibitor. It is intended for research use only and not for patient use.
  • Metabolite of tryptamine
  • Competitive serotonin reuptake inhibitor with a Ki value of 6.1 μM
  • Increases serotonin levels in the brain of rats in vivo studies