Learn More
Cayman Chemical TetrahydroberberIn 10mg
SDP
Catalog No. 502948409
Encompass_Preferred
An isoquinoline alkaloid; inhibits iron- and ascorbate-induced lipid peroxidation in rat liver microsomes (IC50 = 13.7 µM); binds to D2 and 5-HT1A receptors (Kis = 0.83 and 4.2 µM, respectively); inhibits ACh-induced potassium currents in primary rat hippocampal CA1 pyramidal neurons (IC50 = 13 µM); stimulates mouse C12C12 myoblast differentiation
Catalog No. 50-294-8409 Supplier Cayman Chemical Supplier No. 3315710 MG
An isoquinoline alkaloid; inhibits iron- and ascorbate-induced lipid peroxidation in rat liver microsomes (IC50 = 13.7 µM); binds to D2 and 5-HT1A receptors (Kis = 0.83 and 4.2 µM, respectively); inhibits ACh-induced potassium currents in primary rat hippocampal CA1 pyramidal neurons (IC50 = 13 µM); stimulates mouse C12C12 myoblast differentiation