Learn More
Medchemexpress LLC HY-107383 5mg Medchemexpress, Tetrahydrobiopterin CAS:17528-72-2 Purity:>98%
SDP
Catalog No. 501968397
Encompass_Preferred
Medchemexpress, HY-107383 5mg Tetrahydrobiopterin CAS:17528-72-2 Tetrahydrobiopterin is a cofactor of the aromatic amino acid hydroxylases enzymes and also acts as an essential cofactor for all nitric oxide synthase (NOS) isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8397 Supplier Medchemexpress LLC Supplier No. HY1073835MG
Medchemexpress, HY-107383 5mg Tetrahydrobiopterin CAS:17528-72-2 Tetrahydrobiopterin is a cofactor of the aromatic amino acid hydroxylases enzymes and also acts as an essential cofactor for all nitric oxide synthase (NOS) isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.