Learn More
AdipoGen Tetrahydrobostrycin (1 mg)
SDP
Supplier:  AdipoGen AGCN20040M001
Encompass_Preferred
Chemical. CAS: 1072119-07-3 and 1147549-78-7. Formula: C16H20O8. MW: 340.3. Isolated from Fusarium sp. Antibiotic. Antibacterial (weak). Anticancer compound (weak).
Catalog No. 50-205-3388