Learn More
Cayman Chemical TetrahydrocoptIsIn 10mg
SDP
Catalog No. 502947571
Encompass_Preferred
An alkaloid; inhibits LPS-induced NF-κB activation and production of NO, TNF-α, and IL-6 in isolated mouse peritoneal macrophages from 0.001-1 UG/ml; inhibits xylene-induced EAr edema in mice, and it decreases serum levels of TNF-α in a mouse model of LPS-induced septic shock; reduces the severity of ethanol-induced gastric ulcers in mice at 10 or 20 mg/kg
Catalog No. 50-294-7571 Supplier Cayman Chemical Supplier No. 3460310 MG
An alkaloid; inhibits LPS-induced NF-κB activation and production of NO, TNF-α, and IL-6 in isolated mouse peritoneal macrophages from 0.001-1 UG/ml; inhibits xylene-induced EAr edema in mice, and it decreases serum levels of TNF-α in a mouse model of LPS-induced septic shock; reduces the severity of ethanol-induced gastric ulcers in mice at 10 or 20 mg/kg