Learn More
Medchemexpress LLC HY-113114 5mg Medchemexpress, Tetrahydrocortisone CAS:53-05-4 Purity:>98%
SDP
Catalog No. 502080834
Encompass_Preferred
Medchemexpress, HY-113114 5mg Tetrahydrocortisone CAS:53-05-4 Tetrahydrocortisone is a stress-induced hormone. Tetrahydrocortisone is also a urinary metabolite of Cortisone derived from the reduction of Cortisone by 5-reductase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0834 Supplier Medchemexpress LLC Supplier No. HY1131145MG
Medchemexpress, HY-113114 5mg Tetrahydrocortisone CAS:53-05-4 Tetrahydrocortisone is a stress-induced hormone. Tetrahydrocortisone is also a urinary metabolite of Cortisone derived from the reduction of Cortisone by 5-reductase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.