Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrahydrocurcumin | 36062-04-1 | 372.41 | 100 MG
SDP

Catalog No. 5000416807
Encompass_Preferred

Tetrahydrocurcumin is a curcuminoid naturally found in turmeric (Curcuma longa), formed through the reduction of curcumin. This compound is known to inhibit the cytochrome P450 enzymes CYP2C9 and CYP3A4. It is strictly for research purposes and not intended for patient use.

  • High purity of 98.96%.
  • Chemical formula: C21H24O6.
  • Appearance: white to off-white solid.
  • Classified as phenols, polyphenols.
  • Inhibits lipoxygenase at concentrations as low as 1 μM.
  • Cytotoxicity against HEK293 cells with an EC50 of 120 μM.
  • Inhibits STAT3 transcriptional activity in human HaCaT cells.

Catalog No. 50-004-16807 Supplier Medchemexpress LLC Supplier No. HYN0893100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrahydrocurcumin is a curcuminoid naturally found in turmeric (Curcuma longa), formed through the reduction of curcumin. This compound is known to inhibit the cytochrome P450 enzymes CYP2C9 and CYP3A4. It is strictly for research purposes and not intended for patient use.

  • High purity of 98.96%.
  • Chemical formula: C21H24O6.
  • Appearance: white to off-white solid.
  • Classified as phenols, polyphenols.
  • Inhibits lipoxygenase at concentrations as low as 1 μM.
  • Cytotoxicity against HEK293 cells with an EC50 of 120 μM.
  • Inhibits STAT3 transcriptional activity in human HaCaT cells.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.