Learn More
Medchemexpress LLC Tetrahydrocurcumin (Standard) | 36062-04-1 | 372.41 | 50 MG
SDP
Catalog No. 5000416914
Encompass_Preferred
Tetrahydrocurcumin is a curcuminoid found in turmeric (Curcuma longa), produced by the reduction of curcumin. This analytical standard is intended for research and analytical applications, and is known to inhibit CYP2C9 and CYP3A4.
  • Analytical standard
  • Derived from natural sources (turmeric)
  • Intended for research and analytical applications
  • Known to inhibit CYP2C9 and CYP3A4
Catalog No. 50-004-16914 Supplier Medchemexpress LLC Supplier No. HYN089350MG
Tetrahydrocurcumin is a curcuminoid found in turmeric (Curcuma longa), produced by the reduction of curcumin. This analytical standard is intended for research and analytical applications, and is known to inhibit CYP2C9 and CYP3A4.
  • Analytical standard
  • Derived from natural sources (turmeric)
  • Intended for research and analytical applications
  • Known to inhibit CYP2C9 and CYP3A4