Learn More
Medchemexpress LLC Tetrahydrodeoxycorticosterone | 567-03-3 | 99.5% | 334.49 | 1 MG
SDP
Catalog No. 5000437203
Encompass_Preferred
Tetrahydrodeoxycorticosterone is a neurosteroid that acts as a potent positive allosteric modulator (PAM) of the GABAA receptor, possessing potent neuroinhibitory properties.
  • Selectively enhances tonic currents mediated by αβδ receptors.
  • Reduces neuronal excitability in the hippocampus.
  • Enhances tonic currents in thalamocortical neurons.
  • Can induce sedation in animals with hepatic encephalopathy.
Catalog No. 50-004-37203 Supplier Medchemexpress LLC Supplier No. HY1133461MG
Tetrahydrodeoxycorticosterone is a neurosteroid that acts as a potent positive allosteric modulator (PAM) of the GABAA receptor, possessing potent neuroinhibitory properties.
  • Selectively enhances tonic currents mediated by αβδ receptors.
  • Reduces neuronal excitability in the hippocampus.
  • Enhances tonic currents in thalamocortical neurons.
  • Can induce sedation in animals with hepatic encephalopathy.