Learn More
Medchemexpress LLC Tetrahydrodeoxycorticosterone | 567-03-3 | MFCD00067604 | 99.5% | 334.49 g/mol | C21H34O3 | 5 MG
SDP
Catalog No. 5000264845
Encompass_Preferred
Tetrahydrodeoxycorticosterone (THDOC) is an endogenous neurosteroid and a potent positive allosteric modulator of the GABAA receptor. It demonstrates strong neuroinhibitory effects and is used in biochemical and pharmacological research to investigate GABAergic signaling, neurosteroid modulation, and central nervous system activity. Handle according to safety data sheet recommendations and store protected from light.
  • Potent positive allosteric modulator of GABAA receptor.
  • Used to study GABAergic signaling and neurosteroid mechanisms.
  • Exhibits neuroinhibitory, sedative, and anxiolytic activity.
  • Supplied as a research-grade solid with specified purity (≈99.5%).
  • Store protected from light at 4°C; in solvent: -80°C up to 6 months.
Catalog No. 50-002-64845 Supplier Medchemexpress LLC Supplier No. HY1133465MG
Tetrahydrodeoxycorticosterone (THDOC) is an endogenous neurosteroid and a potent positive allosteric modulator of the GABAA receptor. It demonstrates strong neuroinhibitory effects and is used in biochemical and pharmacological research to investigate GABAergic signaling, neurosteroid modulation, and central nervous system activity. Handle according to safety data sheet recommendations and store protected from light.
  • Potent positive allosteric modulator of GABAA receptor.
  • Used to study GABAergic signaling and neurosteroid mechanisms.
  • Exhibits neuroinhibitory, sedative, and anxiolytic activity.
  • Supplied as a research-grade solid with specified purity (≈99.5%).
  • Store protected from light at 4°C; in solvent: -80°C up to 6 months.