Learn More
Cayman Chemical TetrahydroepIberberIn 10mg
SDP
Catalog No. 502911783
Encompass_Preferred
An alkaloid with diverse biological activities; active against the fungus C. albicans (MIC = 8 UG/ml), as well as the bacteria E. coli, P. aeruginosa, P. mirabilis, K. pneumoniae, A. baumannii, S. aureus, and B. subtilis (MICs = 32-64 UG/ml, respectively); inhibits the cytopathogenic effect of PI-3 virus in Vero cells at 16 UG/ml; decreases the viability of HaCaT keratinocytes (IC50 = 150 µM)
Catalog No. 50-291-1783 Supplier Cayman Chemical Supplier No. 3617710 MG
An alkaloid with diverse biological activities; active against the fungus C. albicans (MIC = 8 UG/ml), as well as the bacteria E. coli, P. aeruginosa, P. mirabilis, K. pneumoniae, A. baumannii, S. aureus, and B. subtilis (MICs = 32-64 UG/ml, respectively); inhibits the cytopathogenic effect of PI-3 virus in Vero cells at 16 UG/ml; decreases the viability of HaCaT keratinocytes (IC50 = 150 µM)