Learn More
Medchemexpress LLC Tetrahydrofolic acid | 135-16-0 | C19H23N7O6 | 100 MG
SDP
Catalog No. 5000356040
Encompass_Preferred
Tetrahydrofolic acid is the biologically active vitamin B9 folate derivative. It acts as a donor of one-carbon groups for amino acids, nucleic acids, and lipids, and serves as an acceptor of free formaldehyde, producing 5,10-methylenetetrahydrofolate-Tetrahydrofolic acid.
  • Functions as an endogenous metabolite
  • Induces cytotoxicity in Adh5- and Fanconi anemia (FA)-deficient cells
  • Promotes FANCD2 and ser139-H2AX focus formation in Adh5-/- cells
  • Activates the DNA damage response
Catalog No. 50-003-56040 Supplier Medchemexpress LLC Supplier No. HY14520100MG
Tetrahydrofolic acid is the biologically active vitamin B9 folate derivative. It acts as a donor of one-carbon groups for amino acids, nucleic acids, and lipids, and serves as an acceptor of free formaldehyde, producing 5,10-methylenetetrahydrofolate-Tetrahydrofolic acid.
  • Functions as an endogenous metabolite
  • Induces cytotoxicity in Adh5- and Fanconi anemia (FA)-deficient cells
  • Promotes FANCD2 and ser139-H2AX focus formation in Adh5-/- cells
  • Activates the DNA damage response