Learn More
Medchemexpress LLC HY-14520 25mg Medchemexpress, Tetrahydrofolic acid CAS:135-16-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1452025MG
Encompass_Preferred
Medchemexpress, HY-14520 25mg Tetrahydrofolic acid CAS:135-16-0 Tetrahydrofolic acid (L-5,6,7,8-Tetrahydrofolic acid) is the biologically active vitamin B9 folate derivative. Tetrahydrofolic acid is a donor of one-carbon groups for amino acids, nucleic acids, and lipids. Tetrahydrofolic acid serves as an acceptor of free formaldehyde, producing 5,10-methylenetetrahydrofolate-Tetrahydrofolic acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0538