Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(Tetrahydrofuran-2-yl)methyl]thiourea 98.0+%, TCI America™
SDP

Catalog No. T3413200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3413200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3413200MG Supplier TCI America Supplier No. T3413200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66892-25-9
Molecular Formula C6H12N2OS
Molecular Weight (g/mol) 160.24
MDL Number MFCD00060498
InChI Key DYYSJNQVLWANCC-UHFFFAOYNA-N

Specifications

Melting Point 97°C
Color White
Quantity 200 mg
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 160.24
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material [(Tetrahydrofuran-2-yl)methyl]thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.