Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran, 99.9, 109-99-9, MFCD00005356, 100mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 186562100ML
Encompass
Empirical Formula (Hill Notation) C4H8O, ≥99.9%, Molecular Weight 72.11, bp 65-67 °C (lit.), mp −108 °C (lit.), anhydrous, contains 250 ppm BHT as inhibitor, Synonym: THF
Catalog No. 50-165-6847