Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran anhydrous
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 40175718LP1
Encompass
Tetrahydrofuran (THf) is a saturated cyclic ether with a potential use as a biofuel. Its combustion studies have been investigated. Reports suggest that it is a better promoter than 1 3 dioxolane for CO2-hydrate formation.
Catalog No. 50-290-3075