Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran anhydrous
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1865624X2L
Encompass
Tetrahydrofuran (THf) is a saturated cyclic ether mainly used as an organic solvent. On long term storage it forms organic peroxides. This process can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT removes the free radicals required for the peroxide formation. It constitutes the key fragment of various natural products (polyether antibiotics). THf forms double hydrate with hydrogen sulfide crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies.
Catalog No. 50-290-3395