Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran contains 2
Catalog No. 502899018
Encompass
The product is tetrahydrofuran (THf) and it contains 250ppm BHT (Butylated hydroxytoluene) as an inhibitor. THf is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THf forms double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-1 4-diol is formed as an intermediate during the synthesis of THf. Hot THf is useful for the disSOLUTIONon of polyvinylidene chloride (PVDV).
Catalog No. 50-289-9018 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 873682.5L
The product is tetrahydrofuran (THf) and it contains 250ppm BHT (Butylated hydroxytoluene) as an inhibitor. THf is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THf forms double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-1 4-diol is formed as an intermediate during the synthesis of THf. Hot THf is useful for the disSOLUTIONon of polyvinylidene chloride (PVDV).