Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran contains 2
Catalog No. 502903798
Encompass
Tetrahydrofuran (THf) is a widely used organic solvent. On long term storage it forms organic peroxides. This process can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT removes the free radicals required for the peroxide formation. The effect of THf on the biological systems has been studied.
Catalog No. 50-290-3798 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 36058918LCS
Tetrahydrofuran (THf) is a widely used organic solvent. On long term storage it forms organic peroxides. This process can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT removes the free radicals required for the peroxide formation. The effect of THf on the biological systems has been studied.