Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran-d{8}, 99.5% (Isotopic)

Catalog No. AA3655103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3655103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3655103 Supplier Thermo Scientific Chemicals Supplier No. 03655103
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1693-74-9 | C4H8O | 80.16 g/mol

Deuterated THF is used as a solvent in many organic reactions and in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-74-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 80.16
MDL Number MFCD00044238
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
Synonym tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
PubChem CID 80290
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriooxolane
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 g
Flash Point -17°C (-1°F)
Density 0.985 g/mL
Melting Point -106°C
Boiling Point 66°C
UN Number UN2056
Solubility Information Miscible with water.
Formula Weight 80.16
Percent Purity 99.5%
Chemical Name or Material Tetrahydrofuran-d8, Isotopic
Show More Show Less
Hazard Category H225-H319-H335-H336-H373
Hazard Statement GHS H Statement
H225-H302-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
EINECSNumber 216-898-4
TSCA No
storageNote1 2°C to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.