Learn More
Cambridge Isotope Laboratories Tetrahydrofuran-d8 (d, 99.5%), 1693-74-9, MFCD00044238, 10X1 g
Catalog No. NC9976512
Encompass_Preferred
Chemical Formula: C4D8O, MW: 80.16, 99.5%, Synonyms: THF, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran, THF-d8, 1,4-epoxybutane.
Catalog No. NC9976512 Supplier Cambridge Isotope Laboratories Supplier No. DLM3610X1
Chemical Formula: C4D8O, MW: 80.16, 99.5%, Synonyms: THF, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran, THF-d8, 1,4-epoxybutane.