Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran d8 99.5 at10G
Catalog No. 502897785
Encompass
Tetrahydrofuran-d8 may be used as a solvent in the synthesis of deuteriotris[bis(trimethylsilyl)amido]thorium and deuteriotris[bis(trimethylsiiyl)amido]uranium.
Catalog No. 50-289-7785 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 18431410G
Tetrahydrofuran-d8 may be used as a solvent in the synthesis of deuteriotris[bis(trimethylsilyl)amido]thorium and deuteriotris[bis(trimethylsiiyl)amido]uranium.