Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran d8 99.5 at10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 43772710G
Encompass
Tetrahydrofuran-d8 (THf-d8 TDf) is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. 1H NMR line shape analysis of the exchange rates of proton between methanol and acetic acid dissolved in THf-d8 was reported. It participates as a solvent in the synthesis of deuteriotris[bis(trimethylsilyl)amido]thorium and deuteriotris[bis(trimethylsiiyl)amido]uranium. Dynamic NMR spectroscopic study of 15N 15NAE-dipentadeuterophenylformamidine (DPfA) has been studied in THf-d8.
Catalog No. 50-290-2921