Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran-D8, deuteration degree min 99.5% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1133640010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
10 x 0.75 mL
Packaging:
Glass Ampule
Septum Ampule
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1133640010 10 mL Septum Ampule
M1133640001 1 mL Glass Ampule
M1133640009 10 x 0.75 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1133640010 Supplier MilliporeSigma Supplier No. 1.13364.0010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-74-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 80.16
MDL Number MFCD00044238
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
Synonym tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4
PubChem CID 80290
IUPAC Name (²H₈)oxolane
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Packaging Septum Ampule
Quantity 10 mL
Color Colorless
Odor Ether-like
Assay Percent Range 99.5% min.
Formula Weight 80.16
Chemical Name or Material Tetrahydrofuran-D8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.