Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran inhibitor-
Catalog No. 502898386
Encompass
Tetrahydrofuran (THf) is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THf forms a double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-1 4-diol is formed as an intermediate during the synthesis of THf. Hot THf is useful for the disSOLUTIONon of polyvinylidene chloride (PVDV).
Catalog No. 50-289-8386 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348652L
Tetrahydrofuran (THf) is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THf forms a double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-1 4-diol is formed as an intermediate during the synthesis of THf. Hot THf is useful for the disSOLUTIONon of polyvinylidene chloride (PVDV).