Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019424 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9424 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-9424 Supplier MilliporeSigma Supelco Supplier No. PHR15613X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Tetrahydrofuran is a saturated cyclic ether with a potential use as a biofuel. It is used for the manufacture of polyurethanes.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 109-99-9
Vapor Pressure 114 mmHg (15°C)
Vapor Density 2.5 (vs. air)
Density 0.889 g/mL (at 25°C)
Linear Formula C4H8O
MDL Number MFCD00005356
Melting Point -108°C
Boiling Point 65°C to 67°C
Synonym Tetrahydrofuran; Butylene oxide; Oxolane; Tetramethylene oxide
Quantity 3 x 1.2 mL
Formula Weight 72.11
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.