Learn More
Glen Research Tetrahydrofuran/Pyridine/Acetic Anhydride, 2000 mL, 0.2 micron filtered, Hazardous material: Yes
Catalog No. NC2956555
Encompass_Preferred
This Glen Research product is a high-purity blend of Tetrahydrofuran, Pyridine, and Acetic Anhydride supplied in a 2000 mL volume. It is prepared to exacting specifications for optimal performance in demanding scientific applications, particularly oligonucleotide synthesis. Processed through a 0.2 micron filter during packaging to minimize particulate contamination, this reagent is ideal for ensuring high synthesis efficiency.
  • 2000 mL volume
  • Primary components include Tetrahydrofuran, Pyridine, and Acetic Anhydride
  • Application in oligonucleotide synthesis
  • Processed through a 0.2 micron filter
  • Prepared to exacting specifications
  • Hazardous material
  • Stored at controlled room temperature
Catalog No. NC2956555 Supplier Glen Research Supplier No. 40411062
This Glen Research product is a high-purity blend of Tetrahydrofuran, Pyridine, and Acetic Anhydride supplied in a 2000 mL volume. It is prepared to exacting specifications for optimal performance in demanding scientific applications, particularly oligonucleotide synthesis. Processed through a 0.2 micron filter during packaging to minimize particulate contamination, this reagent is ideal for ensuring high synthesis efficiency.
  • 2000 mL volume
  • Primary components include Tetrahydrofuran, Pyridine, and Acetic Anhydride
  • Application in oligonucleotide synthesis
  • Processed through a 0.2 micron filter
  • Prepared to exacting specifications
  • Hazardous material
  • Stored at controlled room temperature