Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrahydrofuran solution c
Catalog No. 502908032
Encompass
The product is a tetrahydrofuran (THF) SOLUTIONon containing 20%(v/v) 1 2-propanediol. THF a saturated cyclic ester is a water miscible organic solvent. The combustion studies of THF have been reported.
Catalog No. 50-290-8032 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6803034X4L
The product is a tetrahydrofuran (THF) SOLUTIONon containing 20%(v/v) 1 2-propanediol. THF a saturated cyclic ester is a water miscible organic solvent. The combustion studies of THF have been reported.