Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl Chloride 98.0+%, TCI America™
SDP

Catalog No. T080025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T080025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T080025G Supplier TCI America Supplier No. T080025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3003-84-7
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00005370
InChI Key IVJLGIMHHWKRAN-UHFFFAOYSA-N
Synonym tetrahydrofurfuryl chloride, 2-chloromethyl tetrahydrofuran, 2-chloromethyl oxolane, furan, 2-chloromethyl tetrahydro, tetrahydrofurfurylchloride, 2-chloromethyltetrahydrofuran, pubchem7061, acmc-209hcn, enamine_005178
PubChem CID 18150
IUPAC Name 2-(chloromethyl)oxolane
SMILES C1CC(OC1)CCl

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 151°C
Formula Weight 120.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Chloride
RTECSNumber LU0350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.