Learn More
Cayman Chemical TetrahydroharmIn 10mg
SDP
Catalog No. 502972166
Encompass_Preferred
A fluorescent indole alkaloid extracted from B. caapi that inhibits MAO-A and MAO-B with much weaker potency (IC50s = 74 nM and >100 μM, respectively) compared to its companion harmala alkaloids: harmaline (IC50s = 2.5 nM and 25 μM, respectively) and harmine (IC50s = 2 nM and 20 μM, respectively)
Catalog No. 50-297-2166 Supplier Cayman Chemical Supplier No. 1444910 MG
A fluorescent indole alkaloid extracted from B. caapi that inhibits MAO-A and MAO-B with much weaker potency (IC50s = 74 nM and >100 μM, respectively) compared to its companion harmala alkaloids: harmaline (IC50s = 2.5 nM and 25 μM, respectively) and harmine (IC50s = 2 nM and 20 μM, respectively)