Learn More
Cayman Chemical Tetrahydroharmine, Purity: ≥98%, CAS: 17019-01-1, 50 mg
SDP
Catalog No. 502971089
Encompass_Preferred
A fluorescent indole alkaloid extracted from B. caapi that inhibits MAO-A and MAO-B with much weaker potency (IC50s = 74 nM and >100 μM, respectively) compared to its companion harmala alkaloids: harmaline (IC50s = 2.5 nM and 25 μM, respectively) and harmine (IC50s = 2 nM and 20 μM, respectively)
Catalog No. 50-297-1089 Supplier Cayman Chemical Supplier No. 1444950 MG
A fluorescent indole alkaloid extracted from B. caapi that inhibits MAO-A and MAO-B with much weaker potency (IC50s = 74 nM and >100 μM, respectively) compared to its companion harmala alkaloids: harmaline (IC50s = 2.5 nM and 25 μM, respectively) and harmine (IC50s = 2 nM and 20 μM, respectively)