Learn More
Cayman Chemical Tetrahydromagnolol, Purity: ≥98%, CAS: 20601-85-8, 5 mg
SDP
Catalog No. 502955311
Encompass_Preferred
An agonist of CB1 and CB2 and antagonist of GPR55 (KB = 13.3 µM in a β-arrestin translocation assay); a major metabolite of magnolol; binds to the CB1 and CB2 receptors (Kis = 2.26 and 0.416 µM, respectively); inhibits forskolin-induced cAMP accumulation (EC50 = 9.01 and 0.17 µM, respectively); reduces melanin biosynthesis, as well as decreases tyrosinase protein levels by inhibiting tyrosinase maturation and increasing the rate of its degradation
Catalog No. 50-295-5311 Supplier Cayman Chemical Supplier No. 142345 MG
An agonist of CB1 and CB2 and antagonist of GPR55 (KB = 13.3 µM in a β-arrestin translocation assay); a major metabolite of magnolol; binds to the CB1 and CB2 receptors (Kis = 2.26 and 0.416 µM, respectively); inhibits forskolin-induced cAMP accumulation (EC50 = 9.01 and 0.17 µM, respectively); reduces melanin biosynthesis, as well as decreases tyrosinase protein levels by inhibiting tyrosinase maturation and increasing the rate of its degradation