Learn More
Medchemexpress LLC Tetrahydropalmatine hydrochloride | 6024-85-7 | 391.89 | 50 MG
SDP
Catalog No. 5000427790
Encompass_Preferred
Tetrahydropalmatine hydrochloride possesses analgesic effects. It acts through inhibition of amygdaloid release of dopamine to inhibit an epileptic attack in rats.
  • Possesses analgesic effects
  • Inhibits amygdaloid release of dopamine
  • May inhibit epileptic attacks in rats
Catalog No. 50-004-27790 Supplier Medchemexpress LLC Supplier No. HYN0300A50MG
Tetrahydropalmatine hydrochloride possesses analgesic effects. It acts through inhibition of amygdaloid release of dopamine to inhibit an epileptic attack in rats.
  • Possesses analgesic effects
  • Inhibits amygdaloid release of dopamine
  • May inhibit epileptic attacks in rats