Learn More
Medchemexpress LLC Tetrahydropalmatine hydrochloride | 6024-85-7 | 99.9% | 391.89 g/mol | C21H26ClNO4 | 10 MG
SDP
Catalog No. 5000295470
Encompass_Preferred
Tetrahydropalmatine hydrochloride is a research-grade alkaloid salt supplied as a high-purity solid for analytical and biological studies. It is commonly used as an analytical standard and in vitro research reagent where precise dosing and stability are required.
  • High purity (~99.9%) suitable for analytical use.
  • Hydrochloride salt form for improved solubility.
  • Available in small research quantities (10 mg).
  • Solid, white to off-white appearance for easy visual inspection.
  • Store sealed at 4°C, protected from moisture and light.
  • SDS available for handling and safety information.
Catalog No. 50-002-95470 Supplier Medchemexpress LLC Supplier No. HYN0300A10MG
Tetrahydropalmatine hydrochloride is a research-grade alkaloid salt supplied as a high-purity solid for analytical and biological studies. It is commonly used as an analytical standard and in vitro research reagent where precise dosing and stability are required.
  • High purity (~99.9%) suitable for analytical use.
  • Hydrochloride salt form for improved solubility.
  • Available in small research quantities (10 mg).
  • Solid, white to off-white appearance for easy visual inspection.
  • Store sealed at 4°C, protected from moisture and light.
  • SDS available for handling and safety information.