Learn More
Medchemexpress LLC Tetrahydropiperine | 23434-88-0 | MFCD17167032 | 99.4% | 289.37 g·mol-1 | C17H23NO3 | 10 MG
SDP
Catalog No. 5000289845
Encompass_Preferred
Tetrahydropiperine is a research-grade small molecule used to probe inflammatory and survival signaling pathways in cellular and biochemical studies. It acts on NF-κB and MAPK signaling and modulates the PI3K/Akt/mTOR pathway.
  • Selective inhibitor of NF-κB and MAPKs, useful for pathway studies.
  • Modulates PI3K/Akt/mTOR signaling for research on cell survival.
  • High purity solid, suitable for analytical and cellular assays.
  • Available in small research pack sizes for screening and mechanism-of-action work.
  • Molecular weight approximately 289.37 g·mol-1 and formula C17H23NO3.
Catalog No. 50-002-89845 Supplier Medchemexpress LLC Supplier No. HYN420510MG
Tetrahydropiperine is a research-grade small molecule used to probe inflammatory and survival signaling pathways in cellular and biochemical studies. It acts on NF-κB and MAPK signaling and modulates the PI3K/Akt/mTOR pathway.
  • Selective inhibitor of NF-κB and MAPKs, useful for pathway studies.
  • Modulates PI3K/Akt/mTOR signaling for research on cell survival.
  • High purity solid, suitable for analytical and cellular assays.
  • Available in small research pack sizes for screening and mechanism-of-action work.
  • Molecular weight approximately 289.37 g·mol-1 and formula C17H23NO3.