Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydropyran-2-ylmethylamine, 97%

Catalog No. AC453370050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453370050 5 g
AC453370010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453370050 Supplier Thermo Scientific Chemicals Supplier No. 453370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6628-83-7 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6628-83-7
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key NYGCBQKDTGBHSC-UHFFFAOYSA-N
Synonym tetrahydropyran-2-ylmethylamine, 2-aminomethyl tetrahydropyran, tetrahydro-2h-pyran-2-yl methanamine, 2-aminomethyl tetrahydro-2h-pyran, 2h-pyran-2-methanamine, tetrahydro, 2-aminomethyltetrahydropyran, c-tetrahydro-pyran-2-yl-methylamine, pyran, 2-aminomethyl tetrahydro, pyran, 2-aminomethyltetrahydro, oxan-2-yl methanamine
PubChem CID 97924
IUPAC Name oxan-2-ylmethanamine
SMILES C1CCOC(C1)CN

Specifications

Boiling Point 110.0°C (25.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material Tetrahydropyran-2-ylmethylamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.