Learn More
Sigma Aldrich Tetrahydropyran-4-ylmethyl methanesulfonate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Methyl 2,2,2-trichloroacetimidate was used as starting material during the synthesis of novel bibenzimidazole oligomers and polymers
- It was employed as reagent during the synthesis of 2,2α-bisbenzimidazole-5,5α-dicarboxylic acid
Specifications
Specifications
| CAS | 132291-95-3 |
| Molecular Formula | C7H14O4S |
| Linear Formula | C7H14O4S |
| Quantity | 25 g |
| MDL Number | MFCD18785628 |
| Molecular Weight (g/mol) | 194.25 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.