Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrahydrothiophen-3-one | 1003-04-9 | 99.7% | 102.15 | 1 ML
SDP

Catalog No. 5000453943
Encompass_Preferred

Tetrahydrothiophen-3-one is an endogenous metabolite and a pharmaceutical intermediate used to synthesize thiazole compounds. It is found in natural food and is considered safe. This high-purity liquid is ideal for various research applications.

  • Endogenous metabolite and pharmaceutical intermediate.
  • Used to synthesize thiazole compounds.
  • Exists in natural food and has good safety.
  • High purity of 99.72%.
  • Molecular weight of 102.15.
  • Liquid appearance, colorless to light yellow.
  • Soluble in DMSO for in vitro applications.
  • Multiple protocols for in vivo solubility.
  • Recommended storage at 4°C, protected from light.

Catalog No. 50-004-53943 Supplier Medchemexpress LLC Supplier No. HYW01059410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrahydrothiophen-3-one is an endogenous metabolite and a pharmaceutical intermediate used to synthesize thiazole compounds. It is found in natural food and is considered safe. This high-purity liquid is ideal for various research applications.

  • Endogenous metabolite and pharmaceutical intermediate.
  • Used to synthesize thiazole compounds.
  • Exists in natural food and has good safety.
  • High purity of 99.72%.
  • Molecular weight of 102.15.
  • Liquid appearance, colorless to light yellow.
  • Soluble in DMSO for in vitro applications.
  • Multiple protocols for in vivo solubility.
  • Recommended storage at 4°C, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.