Learn More
Abcam Tetrahydrouridine, cytidine deaminase inhibitor, 25MG
Catalog No. 502583330
Encompass_Preferred
MW 248.23 Da, Purity >80%. Competitive cytidine deaminase inhibitor (IC₅₀= 152 μM). Modulates antiproliferative effects. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3330 Supplier Abcam Supplier No. AB14208025MG
MW 248.23 Da, Purity >80%. Competitive cytidine deaminase inhibitor (IC₅₀= 152 μM). Modulates antiproliferative effects. Orally active.
The product is subject to the following: Abcam Restricted Use Statement