Learn More
Medchemexpress LLC Tetrahydrouridine dihydrate | 18771-50-1 | MFCD00063468 | 99.8% | 284.26 g/mol | C9H20N2O8 | 10 MG
SDP
Catalog No. 5000287571
Encompass_Preferred
Tetrahydrouridine dihydrate is the dihydrated form of tetrahydrouridine, a potent competitive inhibitor of cytidine deaminase (CDA). Supplied as a white to off-white solid for research use, it is used to inhibit cytidine deamination, support studies of DNA/RNA synthesis pathways, and sensitize carcinoma cells to cytidine analog chemotherapeutics such as gemcitabine.
  • Potent competitive inhibitor of cytidine deaminase (CDA).
  • High purity (≈99.8%) suitable for research applications.
  • Dihydrate form; molecular weight 284.26 g/mol.
  • White to off-white solid, convenient for analytical and biological assays.
  • Stable when stored as powder at -20°C (long-term storage).
  • Available in small assay sizes for laboratory use.
Catalog No. 50-002-87571 Supplier Medchemexpress LLC Supplier No. HY1534510MG
Tetrahydrouridine dihydrate is the dihydrated form of tetrahydrouridine, a potent competitive inhibitor of cytidine deaminase (CDA). Supplied as a white to off-white solid for research use, it is used to inhibit cytidine deamination, support studies of DNA/RNA synthesis pathways, and sensitize carcinoma cells to cytidine analog chemotherapeutics such as gemcitabine.
  • Potent competitive inhibitor of cytidine deaminase (CDA).
  • High purity (≈99.8%) suitable for research applications.
  • Dihydrate form; molecular weight 284.26 g/mol.
  • White to off-white solid, convenient for analytical and biological assays.
  • Stable when stored as powder at -20°C (long-term storage).
  • Available in small assay sizes for laboratory use.