Learn More
Medchemexpress LLC Tetrahydrouridine dihydrate | C9H20N2O8 | 1 ML
SDP
Catalog No. 5000372665
Encompass_Preferred
Tetrahydrouridine dihydrate (THU dihydrate) is a potent inhibitor of cytidine deaminase (CDA), effectively blocking the enzyme's active site more effectively than intrinsic cytidine. This compound is intended for research use only.
  • Potent inhibitor of cytidine deaminase (CDA)
  • Competitively blocks the enzyme's active site
  • More effective than intrinsic cytidine
  • Suppresses deamination in the catabolism of cytotoxic deoxycytidine analogues
  • Sensitizes some pancreatic and lung carcinoma cells to gemcitabine
  • Inhibits S-phase without inducing apoptosis
Catalog No. 50-003-72665 Supplier Medchemexpress LLC Supplier No. HY1534510MM/1ML
Tetrahydrouridine dihydrate (THU dihydrate) is a potent inhibitor of cytidine deaminase (CDA), effectively blocking the enzyme's active site more effectively than intrinsic cytidine. This compound is intended for research use only.
  • Potent inhibitor of cytidine deaminase (CDA)
  • Competitively blocks the enzyme's active site
  • More effective than intrinsic cytidine
  • Suppresses deamination in the catabolism of cytotoxic deoxycytidine analogues
  • Sensitizes some pancreatic and lung carcinoma cells to gemcitabine
  • Inhibits S-phase without inducing apoptosis