Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydroxy-1,4-benzoquinone Disodium Salt, TCI America™
SDP

Catalog No. T01221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T01221G Supplier TCI America Supplier No. T01221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1887-02-1
Molecular Formula C6H2Na2O6
Molecular Weight (g/mol) 216.056
MDL Number MFCD00036127
InChI Key SOSMJTGSYNGZAV-UHFFFAOYSA-L
Synonym Tetrahydroxyquinone Disodium Salt
PubChem CID 74675
IUPAC Name disodium;4,6-dihydroxy-2,5-dioxocyclohexa-3,6-diene-1,3-diolate
SMILES C1(=C(C(=O)C(=C(C1=O)O)[O-])[O-])O.[Na+].[Na+]

Specifications

Quantity 1 g
Formula Weight 216.06
Physical Form Crystalline Powder
Chemical Name or Material Tetrahydroxy-1,4-benzoquinone Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.