Learn More
Cayman Chemical Tetrahydroxyquinone, Purity: ≥98%, CAS: 319-89-1, 5 g
SDP
Catalog No. 502950374
Encompass_Preferred
A redox-active hydroxyquinone; inhibits HIV-1 proteinase (IC50 = 575 µM) and is active against T. brucei brucei (MIC100 = 100 µg/ml); induces ROS formation and apoptosis in HL-60 leukemia cells at 100 µM
Catalog No. 50-295-0374 Supplier Cayman Chemical Supplier No. 329085 G
A redox-active hydroxyquinone; inhibits HIV-1 proteinase (IC50 = 575 µM) and is active against T. brucei brucei (MIC100 = 100 µg/ml); induces ROS formation and apoptosis in HL-60 leukemia cells at 100 µM