Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydroxyquinone, Solstice

Catalog No. 60047710 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-710 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-710 Supplier Solstice Supplier No. 875105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Titration Indicator (for Ba, SO4)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5676-48-2
Molecular Formula C6H8O8
Molecular Weight (g/mol) 208.122
MDL Number MFCD00001597
InChI Key PTIMJXRLFTZAOV-UHFFFAOYSA-N
PubChem CID 19849282
IUPAC Name 2,3,5,6-tetrahydroxycyclohexa-2,5-diene-1,4-dione;dihydrate
SMILES C1(=C(C(=O)C(=C(C1=O)O)O)O)O.O.O

Specifications

Packaging Glass bottle
UN Number NONH for all modes of transport
Beilstein 1371548
Quantity 5 g
Physical Form Powder
Formula Weight 208.12g/mol
Chemical Name or Material Tetrahydroxyquinone
Hazard Statement P261-P305 + P351 + P338
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.