Learn More
Medchemexpress LLC HY-B1106A 100mg Medchemexpress, Tetrahydroxyquinone (hydrate) CAS:1215458-51-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1106A100MG
Encompass_Preferred
Medchemexpress, HY-B1106A 100mg Tetrahydroxyquinone (hydrate) CAS:1215458-51-7 Tetrahydroxyquinone hydrate (Tetrahydroxy-1,4-benzoquinone hydrate), a primitive anticataract agent, is a redox active benzoquinone. Tetrahydroxyquinone hydrate can take part in a redox cycle with semiquinone radicals, leading to the formation of reactive oxygen species (ROS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2684