Learn More
TARGETMOL CHEMICALS INC Tetrahydroxystilbene-2-O-beta-
SDP
Supplier:  TARGETMOL CHEMICALS INC T49565MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tetrahydroxystilbene-2-O-beta-D-glucoside (EH-201) is a low MW inducer of erythropoietin. Tetrahydroxystilbene-2-O-beta-D-glucoside induces expression of erythropoietin, PPAR-gamma coactivator 1alpha (PGC-1alpha) and haemoglobin in astrocytes and PC12 neuronal-like cells. In vivo, Tetrahydroxystilbene-2-O-beta-D-glucoside treatment restores memory impairment, as assessed by the passive avoidance test, in SD, Abeta and KA mouse models. In the hippocampus of mice given Tetrahydroxystilbene-2-O-beta-D-glucoside in their diet, levels of erythropoietin, PGC-1alpha and haemoglobin were increased. Treatment with Tetrahydroxystilbene-2-O-beta-D-glucoside might be a therapeutic strategy for memory impairment in neurodegenerative disease, physiological ageing or traumatic brain injury. Purity 99.47%
Catalog No. 50-287-8896