Learn More
Apexbio Technology LLC Tetrahydrozoline HCl 522-48-5 5g
SDP
Catalog No. 5000063358
Encompass_Preferred
Tetrahydrozoline HCl (CAS 522-48-5) is an imidazoline-based small molecule that functions primarily as an agonist at alpha-adrenergic receptors Activation of these receptors by tetrahydrozoline elicits vasoconstriction via smooth muscle contraction reducing redness and swelling Due to this mechanism tetrahydrozoline hydrochloride is commonly utilized as a research tool to investigate alpha receptor-mediated physiological and pharmacological responses particularly in cardiovascular and ocular experimental models
Catalog No. 50-000-63358 Supplier Apexbio Technology LLC Supplier No. B13495000
Tetrahydrozoline HCl (CAS 522-48-5) is an imidazoline-based small molecule that functions primarily as an agonist at alpha-adrenergic receptors Activation of these receptors by tetrahydrozoline elicits vasoconstriction via smooth muscle contraction reducing redness and swelling Due to this mechanism tetrahydrozoline hydrochloride is commonly utilized as a research tool to investigate alpha receptor-mediated physiological and pharmacological responses particularly in cardiovascular and ocular experimental models